Product Details

SNP ID
rs7124693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134332142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCGTCTTGGGCTTCCTGGTGCT[C/T]CGCAGGTGAGAGAGAGCTTCGCGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLB1L2 PubMed Links

Gene Details

Gene
GLB1L2
Gene Name
galactosidase beta 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138342.3 269 Silent Mutation CTC,CTT L27L NP_612351.2
XM_006718937.1 269 Silent Mutation CTC,CTT L27L XP_006719000.1

View Full Product Details