Product Details

SNP ID
rs4682859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42822247 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGTAATGGTGGCTTTATCTGAAC[A/G]GTAGAATACTTCGTGATTTTTATTT
Phenotype
MIM: 602648
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACKR2 PubMed Links

Gene Details

Gene
ACKR2
Gene Name
atypical chemokine receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001296.4 Intron NP_001287.2

View Full Product Details