Product Details
- SNP ID
-
rs4343608
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:13822154 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGGGCATCTGAAAGGAGGCAGCCA[C/T]TCTGGAAAACAGTCTGGCAGCTCCT
- Phenotype
-
MIM: 601570
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WNT7A
PubMed Links
Gene Details
- Gene
- WNT7A
- Gene Name
- Wnt family member 7A
View Full Product Details