Product Details

SNP ID
rs887463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:31619114 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACATTACCTTTAATCTGTGTATA[C/T]TAGTCACAACATTTCATTAGTCTTT
Phenotype
MIM: 602987
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC129 PubMed Links

Gene Details

Gene
CCDC129
Gene Name
coiled-coil domain containing 129
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257967.1 Intron NP_001244896.1
NM_001257968.1 Intron NP_001244897.1
NM_194300.3 Intron NP_919276.2
XM_011515213.1 Intron XP_011513515.1
XM_017011871.1 Intron XP_016867360.1
XM_017011872.1 Intron XP_016867361.1
Gene
PDE1C
Gene Name
phosphodiesterase 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191056.3 Intron NP_001177985.1
NM_001191057.3 Intron NP_001177986.1
NM_001191058.3 Intron NP_001177987.2
NM_001191059.3 Intron NP_001177988.1
NM_001322055.1 Intron NP_001308984.1
NM_001322056.1 Intron NP_001308985.1
NM_001322057.1 Intron NP_001308986.1
NM_001322058.1 Intron NP_001308987.1
NM_001322059.1 Intron NP_001308988.1
NM_005020.4 Intron NP_005011.1
XM_017012264.1 Intron XP_016867753.1
XM_017012265.1 Intron XP_016867754.1
XM_017012266.1 Intron XP_016867755.1
XM_017012267.1 Intron XP_016867756.1

View Full Product Details