Product Details

SNP ID
rs61778493
Assay Type
Functionally Tested
NCBI dbSNP Submissions
28
Location
Chr.1:20666394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTGCCCCTGGCACAGGGCTCCA[A/C]AGCCTCTGGTATCCTGGGGCAGTGG
Phenotype
MIM: 602202 MIM: 605037
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DDOST PubMed Links

Gene Details

Gene
DDOST
Gene Name
dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit
There are no transcripts associated with this gene.

Gene
KIF17
Gene Name
kinesin family member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122819.2 Intron NP_001116291.1
NM_001287212.1 Intron NP_001274141.1
NM_020816.3 Intron NP_065867.2
XM_005245950.3 Intron XP_005246007.1
XM_005245951.3 Intron XP_005246008.1
XM_011541837.1 Intron XP_011540139.1
XM_011541838.1 Intron XP_011540140.1
XM_011541839.2 Intron XP_011540141.1
XM_011541840.2 Intron XP_011540142.1
XM_011541841.2 Intron XP_011540143.1
XM_011541842.1 Intron XP_011540144.1
XM_011541843.1 Intron XP_011540145.1
XM_011541844.2 Intron XP_011540146.1
XM_011541845.2 Intron XP_011540147.1
XM_011541846.2 Intron XP_011540148.1
XM_011541847.2 Intron XP_011540149.1

View Full Product Details