Product Details

SNP ID
rs4605124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:31332504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTGGTGTGGCGCCATCTCCAC[C/T]CCCCCCTTCCCCCTCGCTGAAAGAA
Phenotype
MIM: 605328
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLF13 PubMed Links

Gene Details

Gene
KLF13
Gene Name
Kruppel like factor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302461.1 Intron NP_001289390.1
NM_015995.3 Intron NP_057079.2

View Full Product Details