Product Details

SNP ID
rs1242518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17484585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACTGGCTTCCCGGGGATAGTC[C/T]GCATGCAGTTACTGCAGGCCTCTGC
Phenotype
MIM: 609878 MIM: 605550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED9 PubMed Links

Gene Details

Gene
MED9
Gene Name
mediator complex subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018019.2 Intron NP_060489.1
Gene
RASD1
Gene Name
ras related dexamethasone induced 1
There are no transcripts associated with this gene.

View Full Product Details