Product Details

SNP ID
rs1719130
Assay Type
Functionally Tested
NCBI dbSNP Submissions
24
Location
Chr.17:36088900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCAGGGGCCCCCTGCCTATCTC[C/T]GTCTAGAGAGCTTCTCTCAGTGACT
Phenotype
MIM: 182283
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCL3 PubMed Links

Gene Details

Gene
CCL3
Gene Name
C-C motif chemokine ligand 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002983.2 Intron NP_002974.1

View Full Product Details