Product Details
- SNP ID
-
rs1484076
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:50676722 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGCCCTATTTTGGCGGGGGTGGG[C/G]GCGTGGGGGGAGGAGCCCTTGAATA
- Phenotype
-
MIM: 603123
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DOCK3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113251786] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DOCK3
- Gene Name
- dedicator of cytokinesis 3
- Gene
- MIR4787
- Gene Name
- microRNA 4787
There are no transcripts associated with this gene.
View Full Product Details