Product Details

SNP ID
rs1362633
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:50028661 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACTTTTTCTCTGACTCATGGTCTTA[A/G]CACTTACTTGGTTAATGAACCAGTC
Phenotype
MIM: 616869
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CNEP1R1 PubMed Links
Additional Information
For this assay, SNP(s) [rs7201173] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CNEP1R1
Gene Name
CTD nuclear envelope phosphatase 1 regulatory subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281789.1 Intron NP_001268718.1
NM_153261.5 Intron NP_694993.2

View Full Product Details