Product Details

SNP ID
rs968448
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44255944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGGGCCCAGGGTGGGGAAGGGT[A/G]CAGGACATCAGCAGTTGGCCGGTCT
Phenotype
MIM: 606588
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DNMT3L PubMed Links

Gene Details

Gene
DNMT3L
Gene Name
DNA methyltransferase 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013369.3 Intron NP_037501.2
NM_175867.2 Intron NP_787063.1
Gene
LOC105372833
Gene Name
uncharacterized LOC105372833
There are no transcripts associated with this gene.

View Full Product Details