Product Details

SNP ID
rs1130488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184299394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCCGGGGACAAGGACAAAGAAC[A/T]GGAGCTGGTGAGGCGCGACCCCGAG
Phenotype
MIM: 610145 MIM: 606223
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ECE2 PubMed Links

Gene Details

Gene
ECE2
Gene Name
endothelin converting enzyme 2
There are no transcripts associated with this gene.

Gene
PSMD2
Gene Name
proteasome 26S subunit, non-ATPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278708.1 297 Intron NP_001265637.1
NM_001278709.1 297 Intron NP_001265638.1
NM_002808.4 297 Missense Mutation CAG,CTG Q43L NP_002799.3

View Full Product Details