Product Details
- SNP ID
-
rs9722
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:46599326 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTGCTGTCTGCTTTCTTGCATGACC[A/G]TCTCTGTTACAGGAAAGGTTTGGCT
- Phenotype
-
MIM: 176990
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
S100B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116960944] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- S100B
- Gene Name
- S100 calcium binding protein B
View Full Product Details