Product Details

SNP ID
rs1131598
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:133933478 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCACAAAGTCGGTCTCCAATAAA[C/T]ATATTTGTTGATGAATGACCTCCTT
Phenotype
MIM: 601460
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C3orf36 PubMed Links

Gene Details

Gene
C3orf36
Gene Name
chromosome 3 open reading frame 36
There are no transcripts associated with this gene.

Gene
SLCO2A1
Gene Name
solute carrier organic anion transporter family member 2A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005630.2 3441 UTR 3 NP_005621.2
XM_017007075.1 3441 UTR 3 XP_016862564.1
XM_017007076.1 3441 Intron XP_016862565.1
XM_017007077.1 3441 Intron XP_016862566.1

View Full Product Details