Product Details

SNP ID
rs1804736
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:203309264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAATTTTCTGGTTTGTGGGTTGAA[A/G]CAAAGCTGTGAATCACTGCAGGCTG
Phenotype
MIM: 601597
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BTG2 PubMed Links

Gene Details

Gene
BTG2
Gene Name
BTG anti-proliferation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006763.2 2374 UTR 3 NP_006754.1
Gene
LINC01136
Gene Name
long intergenic non-protein coding RNA 1136
There are no transcripts associated with this gene.

View Full Product Details