Product Details

SNP ID
rs1500532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122579273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTTTTTTCTCAATATTATAT[A/G]TGAGGTTTACTCATTCGCTATGTTG
Phenotype
MIM: 613143 MIM: 612066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DTX3L PubMed Links

Gene Details

Gene
DTX3L
Gene Name
deltex E3 ubiquitin ligase 3L
There are no transcripts associated with this gene.

Gene
PARP15
Gene Name
poly(ADP-ribose) polymerase family member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113523.2 Intron NP_001106995.1
NM_001308320.1 Intron NP_001295249.1
NM_001308321.1 Intron NP_001295250.1
NM_152615.2 Intron NP_689828.1
XM_005247159.3 Intron XP_005247216.1
XM_005247160.3 Intron XP_005247217.1
XM_011512475.2 Intron XP_011510777.1
XM_011512476.2 Intron XP_011510778.1
XM_011512477.2 Intron XP_011510779.1
XM_011512478.2 Intron XP_011510780.1
XM_011512479.2 Intron XP_011510781.1
XM_011512480.2 Intron XP_011510782.1
XM_017005791.1 Intron XP_016861280.1
XM_017005792.1 Intron XP_016861281.1

View Full Product Details