Product Details

SNP ID
rs10246939
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:141972804 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACTTGGCATTGCCTGAGATCAGGA[C/T]GGCTGCATGCCCAGAGGGACAAGCT
Phenotype
MIM: 607751
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAS2R38 PubMed Links

Gene Details

Gene
TAS2R38
Gene Name
taste 2 receptor member 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176817.4 970 Missense Mutation ATC,GTC I296V NP_789787.4

View Full Product Details