Product Details
- SNP ID
-
rs662571
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:39597899 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCAGTGAGCCTGGACGTGGATCCT[C/G]CACCAGAGCTCAGCACTGATGACAT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
B3GALT5-AS1
PubMed Links
Gene Details
- Gene
- B3GALT5-AS1
- Gene Name
- B3GALT5 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details