Product Details

SNP ID
rs1543748
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37074281 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCTGGCCAGCTGCTCCGCAGTGT[C/T]GACGCTGCCAGTGTGGCCTCTTCAG
Phenotype
MIM: 605938 MIM: 602259
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PIGP PubMed Links

Gene Details

Gene
PIGP
Gene Name
phosphatidylinositol glycan anchor biosynthesis class P
There are no transcripts associated with this gene.

Gene
TTC3
Gene Name
tetratricopeptide repeat domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320703.1 Intron NP_001307632.1
NM_001320704.1 Intron NP_001307633.1
NM_003316.3 Intron NP_003307.3
XM_005261048.1 Intron XP_005261105.1
XM_005261049.1 Intron XP_005261106.1
XM_005261050.4 Intron XP_005261107.1
XM_005261052.1 Intron XP_005261109.1
XM_005261054.3 Intron XP_005261111.1
XM_005261055.1 Intron XP_005261112.1
XM_005261056.2 Intron XP_005261113.1
XM_011529739.2 Intron XP_011528041.1
XM_011529740.2 Intron XP_011528042.1
XM_011529742.2 Intron XP_011528044.2
XM_017028458.1 Intron XP_016883947.1
XM_017028459.1 Intron XP_016883948.1
XM_017028460.1 Intron XP_016883949.1
XM_017028461.1 Intron XP_016883950.1
XM_017028462.1 Intron XP_016883951.1
XM_017028463.1 Intron XP_016883952.1
XM_017028464.1 Intron XP_016883953.1
XM_017028465.1 Intron XP_016883954.1
XM_017028466.1 Intron XP_016883955.1
XM_017028467.1 Intron XP_016883956.1

View Full Product Details