Product Details
- SNP ID
-
rs10188
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:46161305 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCGGCCCTTTCCCCTCCGGGGGGA[C/T]GCGCAGGAGGCACCGCGGCCCCGGG
- Phenotype
-
MIM: 606806
MIM: 612412
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FTCD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs118185461,rs145916215] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FTCD
- Gene Name
- formimidoyltransferase cyclodeaminase
There are no transcripts associated with this gene.
- Gene
- SPATC1L
- Gene Name
- spermatogenesis and centriole associated 1-like
View Full Product Details