Product Details

SNP ID
rs28730640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:99072902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAAGTAAATACTTTTGAAATTT[C/T]CCTTCCTCTGTATTAAGATTTACTA
Phenotype
MIM: 103710
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADH5 PubMed Links

Gene Details

Gene
ADH5
Gene Name
alcohol dehydrogenase 5 (class III), chi polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000671.4 Intron NP_000662.3

View Full Product Details