Product Details
- SNP ID
-
rs820478
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:123604482 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCCCCTGCTTTTGCTAATTCTGTG[A/G]TAGGAATTACCTTGCCTTTTTCATT
- Phenotype
-
MIM: 600922
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYLK
PubMed Links
Gene Details
- Gene
- MYLK
- Gene Name
- myosin light chain kinase
There are no transcripts associated with this gene.
- Gene
- MYLK-AS1
- Gene Name
- MYLK antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details