Product Details

SNP ID
rs1055137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:123493716 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGAGGATGTGACATGGATTAGAC[A/G]ATGTCGGTAGTACCAAACTATTTCT
Phenotype
MIM: 615939
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HACD2 PubMed Links

Gene Details

Gene
HACD2
Gene Name
3-hydroxyacyl-CoA dehydratase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017005855.1 1768 UTR 3 XP_016861344.1
XM_017005856.1 1768 Intron XP_016861345.1

View Full Product Details