Product Details
- SNP ID
-
rs10758
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:112652438 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTACCTTAGAATGTGGGTATTGCA[A/G]TCTTTATTCAGGATGTTATCAGTCA
- Phenotype
-
MIM: 137570
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ42351
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7571253] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FLJ42351
- Gene Name
- uncharacterized LOC400999
There are no transcripts associated with this gene.
- Gene
- SLC20A1
- Gene Name
- solute carrier family 20 member 1
View Full Product Details