Product Details

SNP ID
rs13124532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:119495290 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGCTCTGTCTCCACAGCAGAGCA[C/T]GGTACCTAACATGTGGCAGGACTCA
Phenotype
MIM: 603310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC645513 PubMed Links

Gene Details

Gene
LOC645513
Gene Name
septin 7 pseudogene
There are no transcripts associated with this gene.

Gene
PDE5A
Gene Name
phosphodiesterase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083.3 5925 UTR 3 NP_001074.2
NM_033430.2 5925 UTR 3 NP_236914.2
NM_033437.3 5925 UTR 3 NP_246273.2
XM_017008791.1 5925 Intron XP_016864280.1

View Full Product Details