Product Details

SNP ID
rs6833114
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:121139453 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCAATCTCAGCTTTACCTGCCCAC[A/C]AAGCAGGTCTGTCTTTCTGCTCTGA
Phenotype
MIM: 608019
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TNIP3 PubMed Links

Gene Details

Gene
TNIP3
Gene Name
TNFAIP3 interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128843.2 Intron NP_001122315.2
NM_001244764.1 Intron NP_001231693.1
NM_024873.5 Intron NP_079149.3
XM_011532256.2 Intron XP_011530558.1
XM_011532257.2 Intron XP_011530559.1
XM_017008619.1 Intron XP_016864108.1
XM_017008620.1 Intron XP_016864109.1
XM_017008621.1 Intron XP_016864110.1
XM_017008622.1 Intron XP_016864111.1
XM_017008623.1 Intron XP_016864112.1
XM_017008624.1 Intron XP_016864113.1
XM_017008625.1 Intron XP_016864114.1

View Full Product Details