Product Details
- SNP ID
-
rs431289
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:18909196 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGAAACAGGAGATTGGGGAAGGCTG[T/C]AGGCACAGCTGGAGCCAGGCTACCT
- Phenotype
-
MIM: 601279
MIM: 606810
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DGCR6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115691424] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DGCR6
- Gene Name
- DiGeorge syndrome critical region gene 6
- Gene
- PRODH
- Gene Name
- proline dehydrogenase 1
There are no transcripts associated with this gene.
View Full Product Details