Product Details

SNP ID
rs1404500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63628145 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTGTGTAAAACATACATACATAT[A/G]TAAACCTACAGTGAACCTGACCAAG
Phenotype
MIM: 609369
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATL3 PubMed Links

Gene Details

Gene
ATL3
Gene Name
atlastin GTPase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290048.1 3093 UTR 3 NP_001276977.1
NM_015459.4 3093 UTR 3 NP_056274.3
XM_006718493.1 3093 UTR 3 XP_006718556.1

View Full Product Details