Product Details

SNP ID
rs1534283
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53299749 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGGGAGCTGCTTATGGGACACCA[C/A]TTCCTGCGCGGCCTCTTAACGCTGC
Phenotype
MIM: 605378 MIM: 611366 MIM: 604143 MIM: 604899
Polymorphism
C/A, Transversion Substitution
Allele Nomenclature
Literature Links
AAAS PubMed Links
Additional Information
For this assay, SNP(s) [rs1534284] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
AAAS
Gene Name
aladin WD repeat nucleoporin
There are no transcripts associated with this gene.

Gene
C12orf10
Gene Name
chromosome 12 open reading frame 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021640.3 64 Missense Mutation CAA,CAC Q4H NP_067653.3
Gene
ESPL1
Gene Name
extra spindle pole bodies like 1, separase
There are no transcripts associated with this gene.

Gene
PFDN5
Gene Name
prefoldin subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002624.3 64 Intron NP_002615.2
NM_145897.2 64 Intron NP_665904.1

View Full Product Details