Product Details

SNP ID
rs1340184
Assay Type
Validated
NCBI dbSNP Submissions
23
Location
Chr.1:178106255 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGGGTAGTGACTGACAGCATAAAA[A/T]ATCTCTTTGCCTGGCTATAATCTAT
Phenotype
MIM: 606136
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RASAL2 PubMed Links

Gene Details

Gene
RASAL2
Gene Name
RAS protein activator like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004841.3 Intron NP_004832.1
NM_170692.2 Intron NP_733793.2
XM_005245622.3 Intron XP_005245679.1
XM_011510166.2 Intron XP_011508468.1
XM_011510167.2 Intron XP_011508469.1
XM_017002849.1 Intron XP_016858338.1
XM_017002850.1 Intron XP_016858339.1
XM_017002851.1 Intron XP_016858340.1
XM_017002852.1 Intron XP_016858341.1
XM_017002853.1 Intron XP_016858342.1
XM_017002854.1 Intron XP_016858343.1
XM_017002855.1 Intron XP_016858344.1

View Full Product Details