Product Details

SNP ID
rs1689509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55909487 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTCAAATGAATTAATCCTTTATG[C/T]GAAGTATTGTCTTAGGGCTTTGAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PYM1 PubMed Links

Gene Details

Gene
PYM1
Gene Name
PYM homolog 1, exon junction complex associated factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143853.1 Intron NP_001137325.1
NM_032345.2 Intron NP_115721.1

View Full Product Details