Product Details

SNP ID
rs1681072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55902181 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTCAAGGCCTCTGCCTCTCCTTT[A/C]TCTTGCTGCTGCCGCCTCTTCTCCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PYM1 PubMed Links

Gene Details

Gene
PYM1
Gene Name
PYM homolog 1, exon junction complex associated factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143853.1 458 Silent Mutation NP_001137325.1
NM_032345.2 458 Silent Mutation NP_115721.1

View Full Product Details