Product Details
- SNP ID
-
rs2417861
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:20690646 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCTAAAAAAAACTAACACAAATTA[C/T]TCAATACAACAAATATCCACAGTGT
- Phenotype
-
MIM: 123805
MIM: 613389
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PDE3A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114641595] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PDE3A
- Gene Name
- phosphodiesterase 3A
There are no transcripts associated with this gene.
- Gene
- SLCO1C1
- Gene Name
- solute carrier organic anion transporter family member 1C1
View Full Product Details