Product Details

SNP ID
rs1800049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1078293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGCACCCCGGTGAAAGTCCCTC[A/G]AGGCTGCGGTGGCGACACCACGTCC
Phenotype
MIM: 614147 MIM: 182455
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1QTNF8 PubMed Links

Gene Details

Gene
C1QTNF8
Gene Name
C1q and tumor necrosis factor related protein 8
There are no transcripts associated with this gene.

Gene
SSTR5
Gene Name
somatostatin receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001053.3 Intron NP_001044.1
NM_001172560.1 Intron NP_001166031.1
Gene
SSTR5-AS1
Gene Name
SSTR5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details