Product Details

SNP ID
rs857031
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:56855389 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGTGCTGAACAGTTAACTTAACC[C/T]CTACTTTCTTATCTGTAAAAGAGCA
Phenotype
MIM: 120950
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8A PubMed Links

Gene Details

Gene
C8A
Gene Name
complement component 8 alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000562.2 Intron NP_000553.1
XM_011542079.2 Intron XP_011540381.1
XM_017002234.1 Intron XP_016857723.1

View Full Product Details