Product Details
- SNP ID
-
rs1342433
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
17
- Location
-
Chr.1:56856044 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAATCCTTTTTGAAAATGAACAAA[A/G]AGTCTTGATGTTTGATGAACTTATC
- Phenotype
-
MIM: 120950
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C8A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115850561] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C8A
- Gene Name
- complement component 8 alpha subunit
View Full Product Details