Product Details

SNP ID
rs232854
Assay Type
Functionally Tested
NCBI dbSNP Submissions
35
Location
Chr.1:58655671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGTCAGGAAATATATCCTAAAG[A/T]ATATTACATTATTCTCCCACAGTGA
Phenotype
MIM: 612176
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
MYSM1 PubMed Links

Gene Details

Gene
MYSM1
Gene Name
Myb like, SWIRM and MPN domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085487.2 6816 UTR 3 NP_001078956.1
XM_006710314.3 6816 UTR 3 XP_006710377.1
XM_011540573.2 6816 UTR 3 XP_011538875.1
XM_011540574.2 6816 UTR 3 XP_011538876.1
XM_017000195.1 6816 UTR 3 XP_016855684.1
XM_017000196.1 6816 UTR 3 XP_016855685.1
XM_017000197.1 6816 UTR 3 XP_016855686.1

View Full Product Details