Product Details

SNP ID
rs232852
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:58656617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTTCGTTAGGTTGGGAAGGAAG[G/T]AAGTTCAAGTCTATAGATCTATAAC
Phenotype
MIM: 612176
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MYSM1 PubMed Links

Gene Details

Gene
MYSM1
Gene Name
Myb like, SWIRM and MPN domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085487.2 5870 UTR 3 NP_001078956.1
XM_006710314.3 5870 UTR 3 XP_006710377.1
XM_011540573.2 5870 UTR 3 XP_011538875.1
XM_011540574.2 5870 UTR 3 XP_011538876.1
XM_017000195.1 5870 UTR 3 XP_016855684.1
XM_017000196.1 5870 UTR 3 XP_016855685.1
XM_017000197.1 5870 UTR 3 XP_016855686.1

View Full Product Details