Product Details

SNP ID
rs1803626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:76657963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGGTCTTGAGTTGGTCCCAGCA[A/C]GTCCATTCTTCCCAGACAGCTCATG
Phenotype
MIM: 137207
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LRRC32 PubMed Links

Gene Details

Gene
LRRC32
Gene Name
leucine rich repeat containing 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128922.1 2414 UTR 3 NP_001122394.1
NM_005512.2 2414 UTR 3 NP_005503.1
XM_005273902.3 2414 UTR 3 XP_005273959.1
XM_005273903.4 2414 UTR 3 XP_005273960.2
XM_011544935.2 2414 UTR 3 XP_011543237.1
XM_017017527.1 2414 UTR 3 XP_016873016.1

View Full Product Details