Product Details

SNP ID
rs3910748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75026311 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACCTACCAATGATGTGTTCTGGG[A/G]AGAAGCTGGCTGGTAGCCCTTCTGT
Phenotype
MIM: 609854 MIM: 613766
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPCDC PubMed Links

Gene Details

Gene
PPCDC
Gene Name
phosphopantothenoylcysteine decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301101.1 Intron NP_001288030.1
NM_001301102.1 Intron NP_001288031.1
NM_001301103.1 Intron NP_001288032.1
NM_001301104.1 Intron NP_001288033.1
NM_001301105.1 Intron NP_001288034.1
NM_021823.4 Intron NP_068595.3
XM_017022461.1 Intron XP_016877950.1
XM_017022462.1 Intron XP_016877951.1
XM_017022463.1 Intron XP_016877952.1
XM_017022464.1 Intron XP_016877953.1
XM_017022465.1 Intron XP_016877954.1
Gene
SCAMP5
Gene Name
secretory carrier membrane protein 5
There are no transcripts associated with this gene.

View Full Product Details