Product Details

SNP ID
rs1063293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53065015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGCTCTGGGGTCTTGCCCTCAC[C/G]CTGGCAGGGAGACAGGCCCCAGAGC
Phenotype
MIM: 607717
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6757 PubMed Links

Gene Details

Gene
MIR6757
Gene Name
microRNA 6757
There are no transcripts associated with this gene.

Gene
SPRYD3
Gene Name
SPRY domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032840.2 2233 UTR 3 NP_116229.1
Gene
TNS2
Gene Name
tensin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015319.2 2233 Intron NP_056134.2
NM_170754.2 2233 Intron NP_736610.2
NM_198316.1 2233 Intron NP_938072.1
XM_006719302.3 2233 Intron XP_006719365.1
XM_006719303.1 2233 Intron XP_006719366.1
XM_011538079.1 2233 Intron XP_011536381.1
XM_017019088.1 2233 Intron XP_016874577.1
XM_017019089.1 2233 Intron XP_016874578.1

View Full Product Details