Product Details

SNP ID
rs966263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34270707 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGATGATGCCTCTTCTTAGCTT[A/T]TGGACCCGTTATAGCTTAATTTATG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0355 PubMed Links

Gene Details

Gene
KIAA0355
Gene Name
KIAA0355
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014686.3 Intron NP_055501.2
XM_005259444.1 Intron XP_005259501.1
XM_017027525.1 Intron XP_016883014.1
XM_017027526.1 Intron XP_016883015.1

View Full Product Details