Product Details

SNP ID
rs1551550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35117641 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCTACAACCTGTGCCCCTATCCA[A/C]CATGCCATCCTGCCCACCCACCCAC
Phenotype
MIM: 604996 MIM: 608303
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FXYD3 PubMed Links

Gene Details

Gene
FXYD3
Gene Name
FXYD domain containing ion transport regulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136007.1 Intron NP_001129479.1
NM_001136008.1 Intron NP_001129480.1
NM_001136009.1 Intron NP_001129481.1
NM_001136010.1 Intron NP_001129482.1
NM_001136011.1 Intron NP_001129483.1
NM_001136012.1 Intron NP_001129484.1
NM_005971.3 Intron NP_005962.1
NM_021910.2 Intron NP_068710.1
XM_005258996.4 Intron XP_005259053.1
XM_011527032.2 Intron XP_011525334.1
XM_017026877.1 Intron XP_016882366.1
XM_017026878.1 Intron XP_016882367.1
Gene
LGI4
Gene Name
leucine rich repeat LGI family member 4
There are no transcripts associated with this gene.

Gene
MIR6887
Gene Name
microRNA 6887
There are no transcripts associated with this gene.

View Full Product Details