Product Details

SNP ID
rs1420671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55659395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTGTATGCAGTGTACTATAAAC[C/T]AGGTTCTTAGGATGCAAAATGGAGC
Phenotype
MIM: 163970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC6A2 PubMed Links

Gene Details

Gene
SLC6A2
Gene Name
solute carrier family 6 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001043.3 Intron NP_001034.1
NM_001172501.1 Intron NP_001165972.1
NM_001172502.1 Intron NP_001165973.1
NM_001172504.1 Intron NP_001165975.1
XM_006721263.2 Intron XP_006721326.1
XM_011523295.1 Intron XP_011521597.1
XM_011523296.1 Intron XP_011521598.1
XM_011523297.2 Intron XP_011521599.1
XM_011523298.1 Intron XP_011521600.1
XM_011523299.2 Intron XP_011521601.1
XM_011523300.2 Intron XP_011521602.1

View Full Product Details