Product Details

SNP ID
rs2008683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36151408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGCCCAGAAACCAGCCACCTC[C/T]TACTCTGGGTCCAGCCCCGCCTCCC
Phenotype
MIM: 114170 MIM: 123995
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPNS1 PubMed Links

Gene Details

Gene
CAPNS1
Gene Name
calpain small subunit 1
There are no transcripts associated with this gene.

Gene
COX7A1
Gene Name
cytochrome c oxidase subunit 7A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001864.3 Intron NP_001855.1

View Full Product Details