Product Details
- SNP ID
-
rs917415
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:2514375 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTTTTCATAGACGAAGCCCCCTCC[A/C]GACCCCCACACTCCATCGTGCATTC
- Phenotype
-
MIM: 604430
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GNG7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112501431] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GNG7
- Gene Name
- G protein subunit gamma 7
- Gene
- LOC101929097
- Gene Name
- uncharacterized LOC101929097
View Full Product Details