Product Details

SNP ID
rs791474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1493044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACTGAGGCACAGGGAGGCAGGG[G/T]CTGCCTCTGTGCCTGTTCTGTCTGA
Phenotype
MIM: 600487 MIM: 609346
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCSK4 PubMed Links

Gene Details

Gene
PCSK4
Gene Name
proprotein convertase subtilisin/kexin type 4
There are no transcripts associated with this gene.

Gene
REEP6
Gene Name
receptor accessory protein 6
There are no transcripts associated with this gene.

View Full Product Details