Product Details

SNP ID
rs1065363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1009920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGCGGGGAGGGCGGGGCCATGGA[A/G]AAGGCACTGCAGGGAGCACCAGGCA
Phenotype
MIM: 606651 MIM: 611011
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIN3B PubMed Links

Gene Details

Gene
GRIN3B
Gene Name
glutamate ionotropic receptor NMDA type subunit 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138690.2 2538 Intron NP_619635.1
XM_017026243.1 2538 Intron XP_016881732.1
Gene
LOC105372235
Gene Name
uncharacterized LOC105372235
There are no transcripts associated with this gene.

Gene
TMEM259
Gene Name
transmembrane protein 259
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033026.1 2538 UTR 3 NP_001028198.1
NM_033420.3 2538 UTR 3 NP_219488.1
XM_005259675.2 2538 UTR 3 XP_005259732.1
XM_005259676.2 2538 UTR 3 XP_005259733.1
XM_005259677.2 2538 UTR 3 XP_005259734.1
XM_005259678.2 2538 UTR 3 XP_005259735.1
XM_017027457.1 2538 UTR 3 XP_016882946.1

View Full Product Details