Product Details

SNP ID
rs1202551
Assay Type
Functionally tested
NCBI dbSNP Submissions
65
Location
Chr.1:230842017 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTGAAGACGTCACATTAAGTAAA[A/G]TAAGTCAGTCACAAAAGGGTAAATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf198 PubMed Links

Gene Details

Gene
C1orf198
Gene Name
chromosome 1 open reading frame 198
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136494.1 Intron NP_001129966.1
NM_001136495.1 Intron NP_001129967.1
NM_032800.2 Intron NP_116189.1
XM_017002599.1 Intron XP_016858088.1

View Full Product Details