Product Details

SNP ID
rs1321258
Assay Type
Validated
NCBI dbSNP Submissions
63
Location
Chr.1:230066281 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GAAGCTTTGCGTGTTGCAAGGGCAT[C/T]ACCAAGGTTACATAGATTATGTTAC
Phenotype
MIM: 602274
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALNT2 PubMed Links

Gene Details

Gene
GALNT2
Gene Name
polypeptide N-acetylgalactosaminyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291866.1 Intron NP_001278795.1
NM_004481.4 Intron NP_004472.1
XM_017000963.1 Intron XP_016856452.1
XM_017000964.1 Intron XP_016856453.1
XM_017000965.1 Intron XP_016856454.1
XM_017000966.1 Intron XP_016856455.1

View Full Product Details